Search
Search
| Gene Symbol: Gmcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010956.2 | NP_001010956.1 | 3 - 4 | 690 | 71 | |
| NM_001033931.1 | NP_001029103.1 | 3 - 4 | 690 | 71 | ||
| XM_006236832.2 | XP_006236894.1 | 3 - 4 | 705 | 71 | ||
| GenBank mRNA | AY862307.1 | - | 3 - 4 | 517 | 71 | |
| AY862308.1 | - | 3 - 4 | 485 | 71 | ||
Set Membership: |
|