Search
Search
| Gene Symbol: Creld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024783.1 | NP_001019954.1 | 5 - 6 | 632 | 63 | |
| XM_006237027.2 | XP_006237089.1 | 4 - 5 | 1119 | 63 | ||
| XM_006237028.2 | XP_006237090.1 | 4 - 5 | 677 | 63 | ||
| GenBank mRNA | BC097951.1 | - | 5 - 6 | 632 | 7442421 | 63 |
Set Membership: |
|