Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01426540_g1
          See other TSNAX GE Assays ›
          Gene Symbol:
          Tsnax
          Gene Name:
          translin-associated factor X
          Gene Aliases:
          Trax
          Chromosome Location:
          Chr.19: 57771539 - 57785567 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01426540_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tsnax
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022262.3 NP_071598.1 5 - 6 579 86
          GenBank mRNA BC081715.1 - 5 - 6 579 7130295 86

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tsnax

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          spermatogenesis
          cell differentiation
          RNA phosphodiester bond hydrolysis, endonucleolytic
          single-stranded DNA binding
          endoribonuclease activity
          A2A adenosine receptor binding
          protein complex binding
          sequence-specific DNA binding
          poly(A) RNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P139806 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P139806 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P139806 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P139806 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12189 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701187642 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700251867 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71549 - -
          Agilent Rat cDNA Microarray (G4105A) 700251867 - -
          Agilent Rat cDNA Microarray (G4105A) 701187642 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10812073 - -
          Affymetrix Rat Gene 1.0 ST Array 10812070 - -
          Affymetrix Rat Gene 1.0 ST Array 10812072 - -
          Affymetrix Rat Gene 1.0 ST Array 10812069 - -
          Affymetrix Rat Gene 1.0 ST Array 10812075 - -
          Affymetrix Rat Gene 1.0 ST Array 10812074 - -
          Affymetrix Rat Gene 1.0 ST Array 10812076 - -
          Affymetrix Rat Gene 1.0 ST Array 10812071 - -
          Affymetrix Rat Gene 1.1 ST Array 10812068 - -
          Rat Genome 230 2.0 Array 1387653_PM_at - -
          Rat Genome 230 2.0 Array 1387653_at - -
          Rat Genome 230 2.0 Array 1398823_at - -
          Rat Genome 230 2.0 Array 1398823_PM_at - -
          Affymetrix HT Rat Focus Array 1387653_PM_at - -
          Affymetrix HT Rat Focus Array 1398823_at - -
          Affymetrix HT Rat Focus Array 1387653_at - -
          Affymetrix HT Rat Focus Array 1398823_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900006955.rn.1 TC1900000844.rn.1 RTA TC
          Rat Expression Set 230 1387653_at - -
          Rat Expression Set 230 1398823_at - -
          Rat Expression Set 230 1398823_PM_at - -
          Rat Expression Set 230 1387653_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10812068 - -
          Rat Genome U34 Set rc_AI232706_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21336471 - -
          ABI Rat Genome Survey Microarray 21853242 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352444 - -
          RatRef-12 Expression BeadChip ILMN_54784 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline