Search
Search
| Gene Symbol: Otud4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191700.1 | NP_001178629.1 | 14 - 15 | 1639 | 65 | |
| XM_006255402.2 | XP_006255464.1 | 14 - 15 | 1978 | 65 | ||
| XM_008772493.1 | XP_008770715.1 | 14 - 15 | 2154 | 65 | ||
| XM_008772494.1 | XP_008770716.1 | 14 - 15 | 1710 | 65 | ||
| XM_008772495.1 | XP_008770717.1 | 15 - 16 | 1758 | 65 | ||
Set Membership: |
|