Search
Search

LOC294154| Gene Symbol: LOC294154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772799.1 | XP_008771021.1 | 4 - 5 | 1055 | 95 | |
| XM_008772800.1 | XP_008771022.1 | 4 - 5 | 1169 | 95 | ||
| XM_008772801.1 | XP_008771023.1 | 4 - 5 | 1594 | 95 | ||
Set Membership: |
|