Search
Search

RGD1306739| Gene Symbol: RGD1306739 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134576.1 | NP_001128048.1 | 6 - 7 | 742 | 129 | |
| XM_006256355.2 | XP_006256417.1 | 6 - 7 | 773 | 129 | ||
| XM_006256356.2 | XP_006256418.1 | 6 - 7 | 748 | 129 | ||
Set Membership: |
|