Search
Search
| Gene Symbol: Anks1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107613.3 | NP_001101083.1 | 12 - 13 | 2178 | 103 | |
| XM_006256193.2 | XP_006256255.1 | 13 - 14 | 2253 | 103 | ||
| XM_006256195.2 | XP_006256257.1 | 12 - 13 | 2190 | 103 | ||
| XM_006256196.2 | XP_006256258.1 | 13 - 14 | 2253 | 103 | ||
| XM_006256197.2 | XP_006256259.1 | 12 - 13 | 2190 | 103 | ||
| XM_008772813.1 | XP_008771035.1 | 13 - 14 | 2253 | 103 | ||
| XM_008772814.1 | XP_008771036.1 | 5 - 6 | 2114 | 103 | ||
| XM_008772815.1 | XP_008771037.1 | 3 - 4 | 691 | 103 | ||
Set Membership: |
|