Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01429443_g1
          See other CUTA GE Assays ›
          Gene Symbol:
          Cuta
          Gene Name:
          cutA divalent cation tolerance homolog (E. coli)
          Gene Aliases:
          CutA1
          Chromosome Location:
          Chr.20: 5532024 - 5533632 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01429443_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cuta
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164706.1 NP_001158178.1 5 - 6 441 63
          NM_001164707.1 NP_001158179.1 4 - 5 494 63
          NM_212494.2 NP_997659.2 5 - 6 494 63
          XM_006256120.2 XP_006256182.1 6 - 7 525 63
          GenBank mRNA BP501235.1 - 4 - 5 508 63
          CB586341.1 - 5 - 6 441 63
          DV717217.1 - 4 - 5 466 63
          FQ223106.1 - 3 - 4 168 63
          FQ229047.1 - 3 - 4 168 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001137.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cuta

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          protein localization
          response to metal ion
          protein homooligomerization
          enzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P150751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226864 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P150751 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P150751 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P150751 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P765298 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10831810 - -
          Affymetrix Rat Gene 1.0 ST Array 10831812 - -
          Affymetrix Rat Gene 1.0 ST Array 10831813 - -
          Affymetrix Rat Gene 1.0 ST Array 10831809 - -
          Affymetrix Rat Gene 1.0 ST Array 10831814 - -
          Affymetrix Rat Gene 1.0 ST Array 10831811 - -
          Affymetrix Rat Gene 1.1 ST Array 10831808 - -
          Rat Genome 230 2.0 Array 1398928_PM_at - -
          Rat Genome 230 2.0 Array 1398928_at - -
          Affymetrix HT Rat Focus Array 1398928_PM_at - -
          Affymetrix HT Rat Focus Array 1398928_at - -
          Affymetrix Rat Clariom S Assay PSR2000009615.rn.1 TC2000001137.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000009614.rn.1 TC2000001137.rn.1 RTA TC
          Rat Expression Set 230 1398928_PM_at - -
          Rat Expression Set 230 1398928_at - -
          Affymetrix Rat Gene 1.0 ST Array 10831808 - -
          Rat Genome U34 Set rc_AA892505_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21370809 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356829 - -
          RatRef-12 Expression BeadChip ILMN_69128 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline