Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01430441_m1
          See other NOX3 GE Assays ›
          Gene Symbol:
          Nox3
          Gene Name:
          NADPH oxidase 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 44550994 - 44615760 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01430441_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nox3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004216.1 NP_001004216.1 11 - 12 1457 83
          GenBank mRNA AY573239.1 - 11 - 12 1457 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004399.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nox3

          Panther Classification:

          Molecular Function -

          oxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          temperature homeostasis
          detection of gravity
          response to gravity
          superoxide anion generation
          otolith development
          oxidation-reduction process
          superoxide-generating NADPH oxidase activity
          oxidoreductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P177595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155883 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173995 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135731 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33410 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85597 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162237 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110365 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100609 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64289 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174409 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P177595 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P177595 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P177595 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P177595 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63395 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42117 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100835 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30145 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10717847 - -
          Affymetrix Rat Gene 1.0 ST Array 10717846 - -
          Affymetrix Rat Gene 1.0 ST Array 10717848 - -
          Affymetrix Rat Gene 1.0 ST Array 10717849 - -
          Affymetrix Rat Gene 1.0 ST Array 10717843 - -
          Affymetrix Rat Gene 1.0 ST Array 10717853 - -
          Affymetrix Rat Gene 1.0 ST Array 10717852 - -
          Affymetrix Rat Gene 1.0 ST Array 10717842 - -
          Affymetrix Rat Gene 1.0 ST Array 10717844 - -
          Affymetrix Rat Gene 1.0 ST Array 10717850 - -
          Affymetrix Rat Gene 1.0 ST Array 10717845 - -
          Affymetrix Rat Gene 1.0 ST Array 10717851 - -
          Affymetrix Rat Gene 1.0 ST Array 10717841 - -
          Affymetrix Rat Gene 1.1 ST Array 10717840 - -
          Affymetrix Rat Clariom S Assay PSR0100035055.rn.1 TC0100004399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100035056.rn.1 TC0100004399.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10717840 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21553140 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376975 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline