Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01430567_m1
          See other VIP GE Assays ›
          Gene Symbol:
          Vip
          Gene Name:
          vasoactive intestinal peptide
          Gene Aliases:
          vip/phi27
          Chromosome Location:
          Chr.1: 42165043 - 42177582 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01430567_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vip
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053991.1 NP_446443.1 3 - 4 373 70
          XM_006227827.2 XP_006227889.1 4 - 5 818 70
          XM_006227828.2 XP_006227890.1 4 - 5 821 70
          GenBank mRNA AB089807.1 - 3 - 4 295 70
          AB089808.1 - 2 - 3 530 70
          AB089809.1 - 3 - 4 390 70
          BC158798.1 - 3 - 4 373 7316540 70
          CB583853.1 - 3 - 4 387 70
          X02341.1 - 2 - 3 208 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000428.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          neuropeptide

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of endothelial cell proliferation
          learning or memory
          regulation of signal transduction
          positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
          regulation of protein localization
          negative regulation of apoptotic process
          negative regulation of potassium ion transport
          positive regulation of protein catabolic process
          positive regulation of vasodilation
          mRNA stabilization
          negative regulation of smooth muscle cell proliferation
          regulation of sensory perception of pain
          positive regulation of penile erection
          prolactin secretion
          receptor binding
          hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P096924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783401 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66521 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P096924 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P096924 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P096924 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P719221 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10702711 - -
          Affymetrix Rat Gene 1.0 ST Array 10702707 - -
          Affymetrix Rat Gene 1.0 ST Array 10702708 - -
          Affymetrix Rat Gene 1.0 ST Array 10702709 - -
          Affymetrix Rat Gene 1.0 ST Array 10702713 - -
          Affymetrix Rat Gene 1.0 ST Array 10702712 - -
          Affymetrix Rat Gene 1.0 ST Array 10702710 - -
          Affymetrix Rat Gene 1.1 ST Array 10702706 - -
          Rat Genome 230 2.0 Array 1377146_PM_at - -
          Rat Genome 230 2.0 Array 1377146_at - -
          Affymetrix HT Rat Focus Array 1377146_at - -
          Affymetrix HT Rat Focus Array 1377146_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100002600.rn.1 TC0100000428.rn.1 RTA TC
          Rat Expression Set 230 1377146_PM_at - -
          Rat Expression Set 230 1377146_at - -
          Affymetrix Rat Gene 1.0 ST Array 10702706 - -
          Rat Genome U34 Set X02341cds_at - -
          Rat Genome U34 Set rc_AI101739_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22202849 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351517 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline