Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01430721_m1
          See other ATP6AP2 GE Assays ›
          Gene Symbol:
          Atp6ap2
          Gene Name:
          ATPase, H+ transporting, lysosomal accessory protein 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 11137889 - 11164854 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01430721_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp6ap2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007091.1 NP_001007092.1 8 - 9 945 124
          GenBank mRNA AB188298.1 - 8 - 9 945 124
          BC079339.1 - 945 7106562 124

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00002694.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          angiotensin maturation
          proteolysis
          biological_process
          rostrocaudal neural tube patterning
          positive regulation of Wnt signaling pathway
          positive regulation of transforming growth factor beta1 production
          regulation of MAPK cascade
          eye pigmentation
          head morphogenesis
          molecular_function
          aspartic-type endopeptidase activity
          receptor activity
          enzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1011008 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1011008 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1011008 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1011008 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P317520 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P504019 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1011008 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45206 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29796 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15079 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10936834 - -
          Affymetrix Rat Gene 1.0 ST Array 10936836 - -
          Affymetrix Rat Gene 1.0 ST Array 10936832 - -
          Affymetrix Rat Gene 1.0 ST Array 10936831 - -
          Affymetrix Rat Gene 1.0 ST Array 10936837 - -
          Affymetrix Rat Gene 1.0 ST Array 10936833 - -
          Affymetrix Rat Gene 1.0 ST Array 10936830 - -
          Affymetrix Rat Gene 1.0 ST Array 10936829 - -
          Affymetrix Rat Gene 1.0 ST Array 10936828 - -
          Affymetrix Rat Gene 1.0 ST Array 10936835 - -
          Affymetrix Rat Gene 1.1 ST Array 10936827 - -
          Rat Genome 230 2.0 Array 1379255_PM_at - -
          Rat Genome 230 2.0 Array 1379255_at - -
          Rat Genome 230 2.0 Array 1396279_at - -
          Rat Genome 230 2.0 Array 1396279_PM_at - -
          Affymetrix HT Rat Focus Array 1379255_at - -
          Affymetrix HT Rat Focus Array 1396279_at - -
          Affymetrix HT Rat Focus Array 1396279_PM_at - -
          Affymetrix HT Rat Focus Array 1379255_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00017301.rn.1 TC0X00002694.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017296.rn.1 TC0X00002694.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017295.rn.1 TC0X00002694.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017297.rn.1 TC0X00002694.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017291.rn.1 TC0X00002694.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017299.rn.1 TC0X00002694.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017300.rn.1 TC0X00002694.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017293.rn.1 TC0X00002694.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017292.rn.1 TC0X00002694.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017298.rn.1 TC0X00002694.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017294.rn.1 TC0X00002694.rn.1 RTA TC
          Rat Expression Set 230 1379255_PM_at - -
          Rat Expression Set 230 1396279_PM_at - -
          Rat Expression Set 230 1379255_at - -
          Rat Expression Set 230 1396279_at - -
          Affymetrix Rat Gene 1.0 ST Array 10936827 - -
          Rat Genome U34 Set rc_AI012226_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22290548 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374034 - -
          RatRef-12 Expression BeadChip ILMN_58795 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline