Search
Search
| Gene Symbol: Dpep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008383.1 | NP_001008384.1 | 6 - 7 | 1037 | 66 | |
| XM_006255517.2 | XP_006255579.1 | 7 - 8 | 1058 | 66 | ||
| XM_008772521.1 | XP_008770743.1 | 7 - 8 | 1255 | 66 | ||
| GenBank mRNA | BC085826.1 | - | 6 - 7 | 1037 | 7131855 | 66 |
Set Membership: |
|