Search
Search
| Gene Symbol: Zfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006255656.2 | XP_006255718.1 | 3 - 4 | 323 | 147 | |
| XM_006255657.2 | XP_006255719.1 | 3 - 4 | 1081 | 147 | ||
| XM_006255660.2 | XP_006255722.1 | 3 - 4 | 1047 | 147 | ||
| XM_006255662.2 | XP_006255724.1 | 3 - 4 | 274 | 147 | ||
| XM_008772583.1 | XP_008770805.1 | 4 - 5 | 1278 | 147 | ||
| XM_008772585.1 | XP_008770807.1 | 4 - 5 | 1227 | 147 | ||
| XM_008772586.1 | XP_008770808.1 | 3 - 4 | 651 | 147 | ||
| XM_574241.6 | XP_574241.4 | 3 - 4 | 290 | 147 | ||
Set Membership: |
|