Search
Search
| Gene Symbol: Lphn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022962.1 | NP_075251.1 | 9 - 10 | 2103 | 67 | |
| GenBank mRNA | AF081144.1 | - | 8 - 9 | 2088 | 67 | |
| AF081145.1 | - | 8 - 9 | 2088 | 67 | ||
| AF081146.1 | - | 9 - 10 | 2103 | 67 | ||
| AF081147.1 | - | 9 - 10 | 2103 | 67 | ||
| AF111099.1 | - | 10 - 11 | 2429 | 67 | ||
| U72487.1 | - | 8 - 9 | 1905 | 67 | ||
| U78105.1 | - | 8 - 9 | 2230 | 67 | ||
Set Membership: |
|