Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01431685_g1
          See other TSNAXIP1 GE Assays ›
          Gene Symbol:
          Tsnaxip1
          Gene Name:
          translin-associated factor X interacting protein 1
          Gene Aliases:
          RGD1565341
          Chromosome Location:
          Chr.19: 37795658 - 37813424 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01431685_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tsnaxip1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109130.1 NP_001102600.1 15 - 16 1946 138
          XM_006255527.2 XP_006255589.1 14 - 15 1915 138
          XM_006255529.2 XP_006255591.1 14 - 15 1776 138
          XM_008772525.1 XP_008770747.1 13 - 14 2605 138

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000492.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P667489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165079 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115673 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161605 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P667489 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P667489 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P667489 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P667489 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47980 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10807368 - -
          Affymetrix Rat Gene 1.0 ST Array 10807372 - -
          Affymetrix Rat Gene 1.0 ST Array 10807379 - -
          Affymetrix Rat Gene 1.0 ST Array 10807380 - -
          Affymetrix Rat Gene 1.0 ST Array 10807383 - -
          Affymetrix Rat Gene 1.0 ST Array 10807378 - -
          Affymetrix Rat Gene 1.0 ST Array 10807375 - -
          Affymetrix Rat Gene 1.0 ST Array 10807381 - -
          Affymetrix Rat Gene 1.0 ST Array 10807373 - -
          Affymetrix Rat Gene 1.0 ST Array 10807371 - -
          Affymetrix Rat Gene 1.0 ST Array 10807382 - -
          Affymetrix Rat Gene 1.0 ST Array 10807369 - -
          Affymetrix Rat Gene 1.0 ST Array 10807377 - -
          Affymetrix Rat Gene 1.0 ST Array 10807370 - -
          Affymetrix Rat Gene 1.0 ST Array 10807374 - -
          Affymetrix Rat Gene 1.0 ST Array 10807376 - -
          Affymetrix Rat Gene 1.1 ST Array 10807367 - -
          Affymetrix Rat Gene 1.1 ST Array 10810703 - -
          Rat Genome 230 2.0 Array 1378664_at - -
          Rat Genome 230 2.0 Array 1391164_at - -
          Rat Genome 230 2.0 Array 1391164_PM_at - -
          Rat Genome 230 2.0 Array 1378664_PM_at - -
          Affymetrix HT Rat Focus Array 1378664_at - -
          Affymetrix HT Rat Focus Array 1391164_PM_at - -
          Affymetrix HT Rat Focus Array 1391164_at - -
          Affymetrix HT Rat Focus Array 1378664_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900004204.rn.1 TC1900000492.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900004205.rn.1 TC1900000492.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900004206.rn.1 TC1900000492.rn.1 RTA TC
          Rat Expression Set 230 1391164_PM_at - -
          Rat Expression Set 230 1378664_at - -
          Rat Expression Set 230 1378664_PM_at - -
          Rat Expression Set 230 1391164_at - -
          Affymetrix Rat Gene 1.0 ST Array 10807367 - -
          Rat Genome U34 Set rc_AI103357_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21050395 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374956 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline