Search
Search
| Gene Symbol: Calb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053988.1 | NP_446440.1 | 1 - 2 | 150 | 104 | |
| XM_008772466.1 | XP_008770688.1 | 2 - 3 | 329 | 104 | ||
| GenBank mRNA | BC087603.1 | - | 1 - 2 | 120 | 7312595 | 104 |
| X66974.1 | - | 1 - 2 | 150 | 104 | ||
Set Membership: |
|