Search
Search
| Gene Symbol: Terf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108448.1 | NP_001101918.1 | 5 - 6 | 746 | 75 | |
| NM_001242355.1 | NP_001229284.1 | 5 - 6 | 709 | 75 | ||
| XM_006255564.2 | XP_006255626.1 | 5 - 6 | 1353 | 75 | ||
| GenBank mRNA | BC160846.1 | - | 5 - 6 | 864 | 8362263 | 75 |
| HQ651915.1 | - | 5 - 6 | 709 | 75 | ||
Set Membership: |
|