Search
Search
| Gene Symbol: Prl6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022176.2 | NP_071512.2 | 4 - 5 | 813 | 156 | |
| GenBank mRNA | BC158563.1 | - | 4 - 5 | 813 | 7443132 | 156 |
| FQ221549.1 | - | 4 - 5 | 830 | 156 | ||
| FQ230053.1 | - | 4 - 5 | 831 | 156 | ||
| M31155.1 | - | 4 - 5 | 772 | 156 | ||
Set Membership: |
|