Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01435139_m1
          See other HIVEP1 GE Assays ›
          Gene Symbol:
          Hivep1
          Gene Name:
          human immunodeficiency virus type I enhancer binding protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 22278103 - 22403362 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01435139_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hivep1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105751.1 NP_001099221.1 9 - 10 7068 85
          GenBank mRNA X54250.1 - 4 - 5 880 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001540.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          negative regulation of transcription, DNA-templated
          RNA polymerase II distal enhancer sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II distal enhancer sequence-specific binding
          nucleic acid binding
          DNA binding
          HMG box domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P164970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83970 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5414 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109700 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45091 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60227 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2527 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79295 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170641 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39145 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2432 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107861 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65014 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18680 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157978 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32444 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164970 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164970 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P164970 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P186454 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P522959 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701932785 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38616 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77831 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14777 - -
          Agilent Rat cDNA Microarray (G4105A) 701932785 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10797867 - -
          Affymetrix Rat Gene 1.0 ST Array 10797870 - -
          Affymetrix Rat Gene 1.0 ST Array 10797869 - -
          Affymetrix Rat Gene 1.0 ST Array 10797868 - -
          Affymetrix Rat Gene 1.0 ST Array 10797866 - -
          Affymetrix Rat Gene 1.0 ST Array 10797865 - -
          Affymetrix Rat Gene 1.0 ST Array 10797864 - -
          Affymetrix Rat Gene 1.1 ST Array 10797863 - -
          Rat Genome 230 2.0 Array 1391560_at - -
          Rat Genome 230 2.0 Array 1391560_PM_at - -
          Affymetrix HT Rat Focus Array 1391560_PM_at - -
          Affymetrix HT Rat Focus Array 1391560_at - -
          Affymetrix Rat Clariom S Assay PSR1700010741.rn.1 TC1700001540.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700010742.rn.1 TC1700001540.rn.1 RTA TC
          Rat Expression Set 230 1391560_at - -
          Rat Expression Set 230 1391560_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10797863 - -
          Rat Genome U34 Set X54250mRNA_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22076426 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358961 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline