Search
Search
| Gene Symbol: Nrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053346.1 | NP_445798.1 | 1 - 2 | 389 | 92 | |
| GenBank mRNA | BC087582.1 | - | 1 - 2 | 326 | 7315687 | 92 |
| U88958.1 | - | 1 - 2 | 389 | 92 | ||
Set Membership: |
|