Search
Search

Mrpl41
RGD1560917| Gene Symbol: Mrpl41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013426.1 | NP_001013444.1 | 1 - 2 | 153 | 75 | |
| GenBank mRNA | BC091293.1 | - | 1 - 2 | 153 | 7307323 | 75 |
| Gene Symbol: RGD1560917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750552.3 | XP_003750600.1 | 1 | 189 | 75 | |
Set Membership: |
|