Search
Search
| Gene Symbol: Ripk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191865.1 | NP_001178794.1 | 9 - 10 | 1256 | 70 | |
| GenBank mRNA | BC107468.1 | - | 7 - 8 | 687 | 70 | |
| FQ230093.1 | - | 3 - 4 | 223 | 70 | ||
Set Membership: |
|