Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01437385_m1
          See other MFSD9 GE Assays ›
          Gene Symbol:
          Mfsd9
          Gene Name:
          major facilitator superfamily domain containing 9
          Gene Aliases:
          RGD1562212
          Chromosome Location:
          Chr.9: 47496977 - 47516464 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01437385_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mfsd9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108215.1 NP_001101685.1 5 - 6 664 91
          XM_006244823.2 XP_006244885.1 5 - 6 656 91
          XM_006244824.2 XP_006244886.1 5 - 6 800 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001893.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Transporter 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          transmembrane transport
          transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P809596 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P015320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595934 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137412 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26740 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16577 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015320 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P809596 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P809596 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015320 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P809596 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P015320 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P319281 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24953 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10927667 - -
          Affymetrix Rat Gene 1.0 ST Array 10927665 - -
          Affymetrix Rat Gene 1.0 ST Array 10927666 - -
          Affymetrix Rat Gene 1.0 ST Array 10927663 - -
          Affymetrix Rat Gene 1.0 ST Array 10927662 - -
          Affymetrix Rat Gene 1.0 ST Array 10927664 - -
          Affymetrix Rat Gene 1.1 ST Array 10927661 - -
          Rat Genome 230 2.0 Array 1374604_at - -
          Rat Genome 230 2.0 Array 1378312_at - -
          Rat Genome 230 2.0 Array 1374604_PM_at - -
          Rat Genome 230 2.0 Array 1378312_PM_at - -
          Affymetrix HT Rat Focus Array 1378312_at - -
          Affymetrix HT Rat Focus Array 1378312_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900015564.rn.1 TC0900001893.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900015563.rn.1 TC0900001893.rn.1 RTA TC
          Rat Expression Set 230 1374604_PM_at - -
          Rat Expression Set 230 1374604_at - -
          Rat Expression Set 230 1378312_PM_at - -
          Rat Expression Set 230 1378312_at - -
          Affymetrix Rat Gene 1.0 ST Array 10927661 - -
          Rat Genome U34 Set rc_AI044116_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21850546 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365814 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline