Search
Search
| Gene Symbol: Mgat4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012225.2 | NP_001012225.1 | 8 - 9 | 1049 | 169 | |
| NM_001160155.1 | NP_001153627.1 | 8 - 9 | 1076 | 169 | ||
| XM_006244840.2 | XP_006244902.1 | 8 - 9 | 1101 | 169 | ||
| XM_008767040.1 | XP_008765262.1 | 7 - 8 | 764 | 169 | ||
| GenBank mRNA | BC088215.1 | - | 7 - 8 | 1007 | 7383944 | 169 |
Set Membership: |
|