Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01437938_m1
          See other PPIL3 GE Assays ›
          Gene Symbol:
          Ppil3
          Gene Name:
          peptidylprolyl isomerase (cyclophilin)-like 3
          Gene Aliases:
          Cyp10l
          Chromosome Location:
          Chr.9: 65315407 - 65329069 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01437938_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppil3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175707.3 NP_783638.1 5 - 6 558 87
          XM_006244964.2 XP_006245026.1 5 - 6 628 87
          GenBank mRNA AF315802.1 - 5 - 6 346 87
          BC087645.1 - 5 - 6 558 7311008 87
          FQ224392.1 - 5 - 6 355 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900002027.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppil3

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          mRNA processing
          protein folding
          RNA splicing
          peptidyl-prolyl cis-trans isomerase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P694171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312975 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79167 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50749 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P694171 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P694171 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P694171 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P694171 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5358 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8194 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55833 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10928244 - -
          Affymetrix Rat Gene 1.0 ST Array 10928247 - -
          Affymetrix Rat Gene 1.0 ST Array 10928249 - -
          Affymetrix Rat Gene 1.0 ST Array 10928250 - -
          Affymetrix Rat Gene 1.0 ST Array 10928243 - -
          Affymetrix Rat Gene 1.0 ST Array 10928251 - -
          Affymetrix Rat Gene 1.0 ST Array 10928246 - -
          Affymetrix Rat Gene 1.0 ST Array 10928248 - -
          Affymetrix Rat Gene 1.0 ST Array 10928245 - -
          Affymetrix Rat Gene 1.1 ST Array 10928242 - -
          Rat Genome 230 2.0 Array 1375686_PM_at - -
          Rat Genome 230 2.0 Array 1375686_at - -
          Affymetrix HT Rat Focus Array 1375686_PM_at - -
          Affymetrix HT Rat Focus Array 1375686_at - -
          Affymetrix Rat Clariom S Assay PSR0900016665.rn.1 TC0900002027.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900016668.rn.1 TC0900002027.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900016666.rn.1 TC0900002027.rn.1 RTA TC
          Rat Expression Set 230 1375686_PM_at - -
          Rat Expression Set 230 1375686_at - -
          Affymetrix Rat Gene 1.0 ST Array 10928242 - -
          Rat Genome U34 Set rc_AA892298_at - -
          Rat Genome U34 Set rc_AA892680_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22220241 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369809 - -
          RatRef-12 Expression BeadChip ILMN_56996 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline