Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01438658_g1
          See other NOXA1 GE Assays ›
          Gene Symbol:
          Noxa1
          Gene Name:
          NADPH oxidase activator 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 2299688 - 2309466 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01438658_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Noxa1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100171.1 NP_001093641.1 11 - 12 1216 127
          XM_006233600.2 XP_006233662.1 11 - 12 1479 127

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002655.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          superoxide metabolic process
          regulation of hydrogen peroxide metabolic process
          positive regulation of catalytic activity
          regulation of respiratory burst
          superoxide-generating NADPH oxidase activator activity
          SH3 domain binding
          enzyme binding
          Rac GTPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P100634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538495 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P100634 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P100634 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P100634 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P516535 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10843320 - -
          Affymetrix Rat Gene 1.0 ST Array 10843326 - -
          Affymetrix Rat Gene 1.0 ST Array 10843329 - -
          Affymetrix Rat Gene 1.0 ST Array 10843318 - -
          Affymetrix Rat Gene 1.0 ST Array 10843325 - -
          Affymetrix Rat Gene 1.0 ST Array 10843328 - -
          Affymetrix Rat Gene 1.0 ST Array 10843330 - -
          Affymetrix Rat Gene 1.0 ST Array 10843327 - -
          Affymetrix Rat Gene 1.0 ST Array 10843322 - -
          Affymetrix Rat Gene 1.0 ST Array 10843323 - -
          Affymetrix Rat Gene 1.0 ST Array 10843321 - -
          Affymetrix Rat Gene 1.0 ST Array 10843319 - -
          Affymetrix Rat Gene 1.1 ST Array 10843317 - -
          Rat Genome 230 2.0 Array 1385375_at - -
          Rat Genome 230 2.0 Array 1385375_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300019694.rn.1 TC0300002655.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10843317 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21331047 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365297 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline