Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01438763_m1
          See other ENG GE Assays ›
          Gene Symbol:
          Eng
          Gene Name:
          endoglin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 11679530 - 11717485 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01438763_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eng
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001010968.2 NP_001010968.1 13 - 14 2010 81
          GenBank mRNA AY562420.1 - 13 - 14 1723 81
          BC105860.1 - 13 - 14 2010 7456529 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000265.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eng

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          chronological cell aging
          angiogenesis
          patterning of blood vessels
          vasculogenesis
          response to hypoxia
          positive regulation of protein phosphorylation
          negative regulation of endothelial cell proliferation
          heart looping
          positive regulation of systemic arterial blood pressure
          cell migration involved in endocardial cushion formation
          regulation of transcription, DNA-templated
          transmembrane receptor protein serine/threonine kinase signaling pathway
          transforming growth factor beta receptor signaling pathway
          heart development
          positive regulation of gene expression
          positive regulation of pathway-restricted SMAD protein phosphorylation
          cell migration
          regulation of transforming growth factor beta receptor signaling pathway
          central nervous system vasculogenesis
          extracellular matrix disassembly
          negative regulation of cell migration
          positive regulation of BMP signaling pathway
          negative regulation of protein autophosphorylation
          response to corticosteroid
          positive regulation of collagen biosynthetic process
          intracellular signal transduction
          response to statin
          wound healing
          positive regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter
          smooth muscle tissue development
          artery morphogenesis
          venous blood vessel morphogenesis
          cell motility
          negative regulation of nitric-oxide synthase activity
          cell chemotaxis
          bone development
          negative regulation of pathway-restricted SMAD protein phosphorylation
          extracellular matrix constituent secretion
          detection of hypoxia
          cellular response to mechanical stimulus
          response to transforming growth factor beta
          transforming growth factor beta-activated receptor activity
          transforming growth factor beta receptor, cytoplasmic mediator activity
          type II transforming growth factor beta receptor binding
          galactose binding
          glycosaminoglycan binding
          type I transforming growth factor beta receptor binding
          BMP binding
          protein homodimerization activity
          transforming growth factor beta binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P474559 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P474559 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P474559 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P474559 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P474559 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10940663 - -
          Affymetrix Rat Gene 1.1 ST Array 10940663 - -
          Rat Genome 230 2.0 Array 1372579_at - -
          Rat Genome 230 2.0 Array 1372579_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300003030.rn.1 TC0300000265.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300003031.rn.1 TC0300000265.rn.1 RTA TC
          Rat Expression Set 230 1372579_at - -
          Rat Expression Set 230 1372579_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10940663 - -
          Rat Genome U34 Set rc_AA997851_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline