Search
Search
| Gene Symbol: Grm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114330.1 | NP_001107802.1 | 1 - 2 | 64 | 71 | |
| NM_017011.1 | NP_058707.1 | 1 - 2 | 64 | 71 | ||
| GenBank mRNA | M61099.1 | - | 1 - 2 | 64 | 71 | |
Set Membership: |
|