Search
Search
| Gene Symbol: Ginm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006227626.2 | XP_006227688.1 | 6 - 7 | 774 | 59 | |
| XM_006227627.2 | XP_006227689.1 | 6 - 7 | 768 | 59 | ||
| XM_341726.7 | XP_341727.4 | 6 - 7 | 772 | 59 | ||
| GenBank mRNA | FQ228838.1 | - | 567 | 59 | ||
Set Membership: |
|