Search
Search
| Gene Symbol: Popdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037369.1 | NP_001032446.1 | 1 - 2 | 144 | 121 | |
| XM_006256599.2 | XP_006256661.1 | 1 - 2 | 225 | 121 | ||
| GenBank mRNA | BC105761.1 | - | 1 - 2 | 144 | 5623862 | 121 |
Set Membership: |
|