Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01441729_g1
          See other SNCB GE Assays ›
          Gene Symbol:
          Sncb
          Gene Name:
          synuclein, beta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 10384472 - 10392776 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01441729_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sncb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080777.2 NP_542955.2 5 - 6 571 95
          XM_006253594.2 XP_006253656.1 6 - 7 772 95
          GenBank mRNA BC087579.1 - 5 - 6 571 7314241 95
          D17764.1 - 5 - 6 427 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000182.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sncb

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          chemical synaptic transmission
          dopamine metabolic process
          negative regulation of neuron apoptotic process
          synapse organization
          calcium ion binding
          alpha-tubulin binding
          beta-tubulin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P030789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742079 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83144 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P030789 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P030789 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P030789 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P254957 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92962 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87845 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10794137 - -
          Affymetrix Rat Gene 1.0 ST Array 10794138 - -
          Affymetrix Rat Gene 1.0 ST Array 10794142 - -
          Affymetrix Rat Gene 1.0 ST Array 10794139 - -
          Affymetrix Rat Gene 1.0 ST Array 10794140 - -
          Affymetrix Rat Gene 1.0 ST Array 10794141 - -
          Affymetrix Rat Gene 1.1 ST Array 10794136 - -
          Rat Genome 230 2.0 Array 1369924_at - -
          Rat Genome 230 2.0 Array 1371568_PM_at - -
          Rat Genome 230 2.0 Array 1369924_PM_at - -
          Rat Genome 230 2.0 Array 1371568_at - -
          Affymetrix HT Rat Focus Array 1371568_at - -
          Affymetrix HT Rat Focus Array 1369924_PM_at - -
          Affymetrix HT Rat Focus Array 1369924_at - -
          Affymetrix HT Rat Focus Array 1371568_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700001507.rn.1 TC1700000182.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700001509.rn.1 TC1700000182.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700001508.rn.1 TC1700000182.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700001506.rn.1 TC1700000182.rn.1 RTA TC
          Rat Expression Set 230 1369924_at - -
          Rat Expression Set 230 1371568_PM_at - -
          Rat Expression Set 230 1369924_PM_at - -
          Rat Expression Set 230 1371568_at - -
          Affymetrix Rat Gene 1.0 ST Array 10794136 - -
          Rat Genome U34 Set D17764_at - -
          Rat Genome U34 Set rc_AI145381_at - -
          Rat Genome U34 Set D17764_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21846813 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354192 - -
          RatRef-12 Expression BeadChip ILMN_65288 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline