Search
Search
| Gene Symbol: H2afy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017182.2 | NP_058878.1 | 2 - 3 | 359 | 117 | |
| XM_006253573.2 | XP_006253635.1 | 2 - 3 | 394 | 117 | ||
| XM_006253574.2 | XP_006253636.1 | 2 - 3 | 484 | 117 | ||
| XM_006253575.2 | XP_006253637.1 | 2 - 3 | 394 | 117 | ||
| XM_006253576.2 | XP_006253638.1 | 2 - 3 | 394 | 117 | ||
| XM_006253577.2 | XP_006253639.1 | 2 - 3 | 394 | 117 | ||
| GenBank mRNA | BC089093.1 | - | 2 - 3 | 359 | 7126329 | 117 |
| M99065.2 | - | 1 - 2 | 209 | 117 | ||
| U79139.1 | - | 2 - 3 | 304 | 117 | ||
Set Membership: |
|