Search
Search
| Gene Symbol: Golm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001056825.4 | XP_001056825.3 | 6 - 7 | 705 | 83 | |
| XM_006253546.2 | XP_006253608.1 | 6 - 7 | 985 | 83 | ||
| XM_006253547.2 | XP_006253609.1 | 6 - 7 | 986 | 83 | ||
Set Membership: |
|