Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01442035_m1
          See other ARL10 GE Assays ›
          Gene Symbol:
          Arl10
          Gene Name:
          ADP-ribosylation factor-like 10
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 10567064 - 10575535 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01442035_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arl10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207165.1 NP_997048.1 3 - 4 613 73
          XM_006253629.1 XP_006253691.1 3 - 4 661 73
          GenBank mRNA FQ210279.1 - 3 - 4 606 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001350.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arl10

          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P18324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486108 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18324 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18324 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P18324 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18324 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68236 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10797435 - -
          Affymetrix Rat Gene 1.0 ST Array 10797440 - -
          Affymetrix Rat Gene 1.0 ST Array 10797439 - -
          Affymetrix Rat Gene 1.0 ST Array 10797437 - -
          Affymetrix Rat Gene 1.0 ST Array 10797438 - -
          Affymetrix Rat Gene 1.1 ST Array 10797434 - -
          Rat Genome 230 2.0 Array 1397276_PM_at - -
          Rat Genome 230 2.0 Array 1397276_at - -
          Affymetrix HT Rat Focus Array 1397276_at - -
          Affymetrix HT Rat Focus Array 1397276_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700009473.rn.1 TC1700001350.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700009474.rn.1 TC1700001350.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700009472.rn.1 TC1700001350.rn.1 RTA TC
          Rat Expression Set 230 1397276_PM_at - -
          Rat Expression Set 230 1397276_at - -
          Affymetrix Rat Gene 1.0 ST Array 10797434 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21874382 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361239 - -
          RatRef-12 Expression BeadChip ILMN_60048 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline