Search
Search
| Gene Symbol: Chpf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106574.1 | NP_001100044.1 | 1 - 2 | 1583 | 97 | |
| XM_006235890.2 | XP_006235952.1 | 2 - 3 | 740 | 97 | ||
| XM_006235891.2 | XP_006235953.1 | 1 - 2 | 88 | 97 | ||
| XM_008762633.1 | XP_008760855.1 | 2 - 3 | 762 | 97 | ||
Set Membership: |
|