Search
Search

Smarcd3| Gene Symbol: Smarcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011966.1 | NP_001011966.1 | 4 - 5 | 434 | 141 | |
| XM_006235883.2 | XP_006235945.1 | 3 - 4 | 495 | 141 | ||
| XM_006235884.2 | XP_006235946.1 | 3 - 4 | 495 | 141 | ||
| XM_006235885.2 | XP_006235947.1 | 3 - 4 | 495 | 141 | ||
| XM_006235886.2 | XP_006235948.1 | 4 - 5 | 482 | 141 | ||
| XM_006235887.2 | XP_006235949.1 | 4 - 5 | 482 | 141 | ||
| XM_006235888.2 | XP_006235950.1 | 3 - 4 | 495 | 141 | ||
| XM_006235889.2 | XP_006235951.1 | 3 - 4 | 495 | 141 | ||
| GenBank mRNA | BC085349.1 | - | 4 - 5 | 434 | 7107899 | 141 |
Set Membership: |
|