Search
Search
| Gene Symbol: Gatsl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025128.1 | NP_001020299.1 | 7 - 8 | 884 | 132 | |
| XM_006251335.2 | XP_006251397.1 | 7 - 8 | 952 | 132 | ||
| XM_006251336.2 | XP_006251398.1 | 7 - 8 | 952 | 132 | ||
| GenBank mRNA | BC091274.1 | - | 7 - 8 | 884 | 7383249 | 132 |
Set Membership: |
|