Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01443437_g1
          See other NELFA GE Assays ›
          Gene Symbol:
          Nelfa
          Gene Name:
          negative elongation factor complex member A
          Gene Aliases:
          Whsc2
          Chromosome Location:
          Chr.14: 82093837 - 82117913 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01443437_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nelfa
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008339.1 NP_001008340.1 10 - 11 1467 119
          GenBank mRNA BC085948.1 - 10 - 11 1467 7130899 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000839.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nelfa

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription elongation from RNA polymerase II promoter
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of histone H3-K4 methylation
          chromatin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1024965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185630 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32497 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90463 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1024965 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1024965 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1024965 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P506826 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1024965 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100398 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6289 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37332 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10773564 - -
          Affymetrix Rat Gene 1.0 ST Array 10773563 - -
          Affymetrix Rat Gene 1.0 ST Array 10773567 - -
          Affymetrix Rat Gene 1.0 ST Array 10773560 - -
          Affymetrix Rat Gene 1.0 ST Array 10773565 - -
          Affymetrix Rat Gene 1.0 ST Array 10773569 - -
          Affymetrix Rat Gene 1.0 ST Array 10773570 - -
          Affymetrix Rat Gene 1.0 ST Array 10773566 - -
          Affymetrix Rat Gene 1.0 ST Array 10773568 - -
          Affymetrix Rat Gene 1.0 ST Array 10773562 - -
          Affymetrix Rat Gene 1.0 ST Array 10773561 - -
          Affymetrix Rat Gene 1.1 ST Array 10773559 - -
          Rat Genome 230 2.0 Array 1389175_PM_at - -
          Rat Genome 230 2.0 Array 1389175_at - -
          Affymetrix HT Rat Focus Array 1389175_PM_at - -
          Affymetrix HT Rat Focus Array 1389175_at - -
          Affymetrix Rat Clariom S Assay PSR1400005869.rn.1 TC1400000839.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400005870.rn.1 TC1400000839.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400005871.rn.1 TC1400000839.rn.1 RTA TC
          Rat Expression Set 230 1389175_PM_at - -
          Rat Expression Set 230 1389175_at - -
          Affymetrix Rat Gene 1.0 ST Array 10773559 - -
          Rat Genome U34 Set rc_AA956953_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22064093 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349806 - -
          RatRef-12 Expression BeadChip ILMN_49970 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline