Search
Search

Pyroxd2| Gene Symbol: Pyroxd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004261.1 | NP_001004261.1 | 1 - 2 | 130 | 101 | |
| XM_006231394.2 | XP_006231456.1 | 1 - 2 | 401 | 101 | ||
| XM_006231396.2 | XP_006231458.1 | 1 - 2 | 402 | 101 | ||
| XM_008760424.1 | XP_008758646.1 | 1 - 2 | 404 | 101 | ||
| XM_008760425.1 | XP_008758647.1 | 1 - 2 | 136 | 132 | ||
| GenBank mRNA | BC079368.1 | - | 1 - 2 | 130 | 7109628 | 101 |
Set Membership: |
|