Search
Search
| Gene Symbol: Crtac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134401.2 | NP_599228.2 | 7 - 8 | 1089 | 106 | |
| XM_006231385.2 | XP_006231447.1 | 7 - 8 | 1151 | 106 | ||
| GenBank mRNA | BC089944.1 | - | 5 - 6 | 620 | 106 | |
Set Membership: |
|