Search
Search
| Gene Symbol: Cadm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012201.1 | NP_001012201.1 | 7 - 8 | 1160 | 88 | |
| XM_006242944.2 | XP_006243006.1 | 7 - 8 | 1159 | 88 | ||
| GenBank mRNA | BC078966.1 | - | 7 - 8 | 1160 | 7111523 | 88 |
Set Membership: |
|