Search
Search
| Gene Symbol: Cadm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006242944.2 | XP_006243006.1 | 9 - 10 | 1287 | 59 | |
| XM_006242947.2 | XP_006243009.1 | 8 - 9 | 1197 | 59 | ||
| GenBank mRNA | AB114443.1 | - | 8 - 9 | 1087 | 59 | |
Set Membership: |
|