Search
Search
| Gene Symbol: Tinag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005549.1 | NP_001005549.1 | 7 - 8 | 1133 | 111 | |
| GenBank mRNA | BC081887.1 | - | 7 - 8 | 1133 | 7109073 | 111 |
Set Membership: |
|