Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01445234_g1
          See other SYNCRIP GE Assays ›
          Gene Symbol:
          Syncrip
          Gene Name:
          synaptotagmin binding, cytoplasmic RNA interacting protein
          Gene Aliases:
          Ab2-339
          Chromosome Location:
          Chr.8: 96100692 - 96266342 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01445234_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Syncrip
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001047916.1 NP_001041381.1 5 - 6 571 137
          XM_006243513.2 XP_006243575.1 1309 137
          XM_006243515.1 XP_006243577.1 1264 137
          XM_006243516.1 XP_006243578.1 1264 137
          XM_006243517.2 XP_006243579.1 4 - 5 536 137
          XM_006243518.2 XP_006243580.1 6 - 7 760 137
          XM_006243519.2 XP_006243581.1 6 - 7 760 137
          GenBank mRNA AY325202.1 - 5 - 6 571 137

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          mRNA processing
          RNA splicing
          negative regulation of translation
          CRD-mediated mRNA stabilization
          cellular response to interferon-gamma
          nucleotide binding
          nucleic acid binding
          RNA binding
          poly(A) binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P135557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389448 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119842 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70968 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78722 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130588 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43632 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14159 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16829 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117520 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139103 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116520 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P135557 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P135557 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P135557 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P382696 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P319236 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P263669 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P776823 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P539947 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1026560 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P289828 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13419 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93681 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3595 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8235 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97053 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10919216 - -
          Affymetrix Rat Gene 1.0 ST Array 10919218 - -
          Affymetrix Rat Gene 1.0 ST Array 10919221 - -
          Affymetrix Rat Gene 1.0 ST Array 10919217 - -
          Affymetrix Rat Gene 1.0 ST Array 10919219 - -
          Affymetrix Rat Gene 1.0 ST Array 10919220 - -
          Affymetrix Rat Gene 1.1 ST Array 10919214 - -
          Rat Genome 230 2.0 Array 1395758_PM_at - -
          Rat Genome 230 2.0 Array 1390489_PM_at - -
          Rat Genome 230 2.0 Array 1390489_at - -
          Rat Genome 230 2.0 Array 1383051_at - -
          Rat Genome 230 2.0 Array 1383051_PM_at - -
          Rat Genome 230 2.0 Array 1393265_at - -
          Rat Genome 230 2.0 Array 1382923_at - -
          Rat Genome 230 2.0 Array 1395758_at - -
          Rat Genome 230 2.0 Array 1393265_PM_at - -
          Rat Genome 230 2.0 Array 1382923_PM_at - -
          Affymetrix HT Rat Focus Array 1395758_at - -
          Affymetrix HT Rat Focus Array 1383051_at - -
          Affymetrix HT Rat Focus Array 1382923_PM_at - -
          Affymetrix HT Rat Focus Array 1393265_PM_at - -
          Affymetrix HT Rat Focus Array 1395758_PM_at - -
          Affymetrix HT Rat Focus Array 1382923_at - -
          Affymetrix HT Rat Focus Array 1393265_at - -
          Affymetrix HT Rat Focus Array 1383051_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800030553.rn.1 TC0800003690.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800030548.rn.1 TC0800003690.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800030545.rn.1 TC0800003690.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800030546.rn.1 TC0800003690.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800030551.rn.1 TC0800003690.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800030547.rn.1 TC0800003690.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800030550.rn.1 TC0800003690.rn.1 RTA TC
          Rat Expression Set 230 1383051_PM_at - -
          Rat Expression Set 230 1390489_at - -
          Rat Expression Set 230 1390489_PM_at - -
          Rat Expression Set 230 1383051_at - -
          Rat Expression Set 230 1382923_PM_at - -
          Rat Expression Set 230 1393265_at - -
          Rat Expression Set 230 1382923_at - -
          Rat Expression Set 230 1393265_PM_at - -
          Rat Expression Set 230 1395758_PM_at - -
          Rat Expression Set 230 1395758_at - -
          Affymetrix Rat Gene 1.0 ST Array 10919214 - -
          Rat Genome U34 Set rc_AA924802_at - -
          Rat Genome U34 Set rc_AI072424_at - -
          Rat Genome U34 Set rc_AI170816_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21516915 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350584 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline