Search
Search
| Gene Symbol: Csrnp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108786.1 | NP_001102256.1 | 3 - 4 | 562 | 63 | |
| XM_006244119.2 | XP_006244181.1 | 3 - 4 | 1229 | 63 | ||
| XM_006244120.2 | XP_006244182.1 | 3 - 4 | 834 | 63 | ||
| XM_006244121.1 | XP_006244183.1 | 3 - 4 | 2985 | 63 | ||
| XM_006244122.2 | XP_006244184.1 | 3 - 4 | 579 | 63 | ||
| XM_008766688.1 | XP_008764910.1 | 2 - 3 | 3902 | 63 | ||
| XM_008766689.1 | XP_008764911.1 | 3 - 4 | 2846 | 63 | ||
Set Membership: |
|