Search
Search
| Gene Symbol: Als2cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244011.2 | XP_006244073.1 | 23 - 24 | 3675 | 62 | |
| XM_006244013.2 | XP_006244075.1 | 24 - 25 | 3507 | 62 | ||
| XM_006244014.2 | XP_006244076.1 | 23 - 24 | 3706 | 62 | ||
| XM_006244015.2 | XP_006244077.1 | 24 - 25 | 3762 | 62 | ||
| XM_006244016.2 | XP_006244078.1 | 24 - 25 | 3759 | 62 | ||
| XM_006244018.2 | XP_006244080.1 | 23 - 24 | 3882 | 62 | ||
| XM_008766654.1 | XP_008764876.1 | 23 - 24 | 3709 | 62 | ||
| XM_008766655.1 | XP_008764877.1 | 13 - 14 | 1600 | 62 | ||
| XM_236654.8 | XP_236654.6 | 23 - 24 | 3916 | 62 | ||
Set Membership: |
|