Search
Search
| Gene Symbol: Klhl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244005.1 | XP_006244067.1 | 4 - 5 | 671 | 55 | |
| XM_006244006.2 | XP_006244068.1 | 4 - 5 | 1327 | 55 | ||
| XM_006244007.1 | XP_006244069.1 | 4 - 5 | 485 | 55 | ||
| XM_236647.7 | XP_236647.6 | 4 - 5 | 671 | 55 | ||
Set Membership: |
|