Search
Search
| Gene Symbol: Azi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025705.1 | NP_001020876.1 | 3 - 4 | 711 | 69 | |
| XM_006244032.2 | XP_006244094.1 | 4 - 5 | 1582 | 69 | ||
| GenBank mRNA | BC098680.1 | - | 3 - 4 | 711 | 7384893 | 69 |
Set Membership: |
|