Search
Search
| Gene Symbol: Nktr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244160.2 | XP_006244222.1 | 9 - 10 | 4123 | 71 | |
| XM_006244164.2 | XP_006244226.1 | 9 - 10 | 3769 | 71 | ||
| XM_008766704.1 | XP_008764926.1 | 10 - 11 | 4189 | 71 | ||
| XM_008766705.1 | XP_008764927.1 | 10 - 11 | 4189 | 71 | ||
| XM_008766706.1 | XP_008764928.1 | 11 - 12 | 4217 | 71 | ||
| XM_008766707.1 | XP_008764929.1 | 8 - 9 | 6461 | 71 | ||
Set Membership: |
|