Search
Search
| Gene Symbol: Acy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005383.1 | NP_001005383.1 | 8 - 9 | 661 | 75 | |
| XM_006243715.2 | XP_006243777.1 | 9 - 10 | 963 | 75 | ||
| XM_006243716.2 | XP_006243778.1 | 9 - 10 | 967 | 75 | ||
| GenBank mRNA | AY580164.1 | - | 8 - 9 | 661 | 75 | |
| BC078930.1 | - | 9 - 10 | 724 | 7107165 | 75 | |
| FQ232544.1 | - | 9 - 10 | 819 | 75 | ||
Set Membership: |
|